missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Type 5/6 Classic Light Chemical Protection Suit

Product Code. 17707539 Shop All Uvex Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 17707539

Brand: Uvex™ 1759510

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight and breathable chemical protection suit offers reliable protection with first-class comfort.

  • PP spunbond non-woven fabric with PE film laminated, ∼55 g/m²
  • Extremely lightweight and breathable material guarantees an outstanding combination of comfort and safety
  • Soft non-woven material lining
  • Inner overlock seam for outstanding protection against particles and liquid splashes
  • Middle finger loops prevent the sleeve from riding up
  • Optimum fit for a high level of wearer comfort
  • Secure, comfortable hems thanks to elasticated bands in the hood, cuffs and ankles
  • Particle-tight with limited spray-tightness
  • Anti-static properties
  • Protection against infectious diseases
  • Free of substances that interfere with wetting agents
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Open
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance Chemical protection/PPE category III, EN ISO 13982-1:2004+A1:2010 (type 5), EN 13034:2005+A1:2010 (type 6), EN 1149-5:2008, EN 14126:2003+AC:2004, EN 1073-2:2002
For Use With (Application) Controlled Environments
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.