Learn More
Saint Gobain Life Sciences Tygon™ E-3603 Bio-based Tubing
Click to view available options
Diameter (English) Inner:
0.08 in.
Unit Size:
15m
Description
- The new bio-based formulation delivers the same excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Specifications
| Certifications/Compliance | According FDA |
| Diameter (English) Inner | 0.08 in. |
| Diameter (Metric) Inner | 2 mm |
| Material | Tygon E-3603 |
| Diameter (English) Outer | 0.16 in. |
| Diameter (Metric) Outer | 4 mm |
| Overall Length (English) | 49.2 ft. |
| Overall Length (Metric) | 15 m |
| Product Type | Tubing |