Learn More
Heidolph™ Tygon (Standard) Tubing
Shop All Heidolph Instruments Products
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.7 mm
3.1 mm
7.9 mm
Unit Size:
1m
Each
Description
- Non-toxic.
- Non-oxidizing.
- Good resistance to acids, bases and inorganic media.
- Very low gas permeability.
- Good performance life.
Specifications
Specifications
| Certifications/Compliance | FDA (21 CFR 177.2601) and GLP |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 3.1 mm |
| Material | Thermoplastic |
| Diameter (Metric) Outer | 6.3 mm |
| Thickness (Metric) | 1.6 mm |
| For Use With | Pump |
| For Use With (Application) | Non-Toxic, Non-Oxidizing, Good Resistance to Acids, Bases and Inorganic Media, Very Low Gas Permeability, Good Performance Life. |
| Min. Operating Temperature (Metric) | -50°C |
| Sterility | Non-sterile |
| Show More |