Learn More
Ismatec™ Tygon ST Extension Tubing
Brand: Ismatec™ SC0047T
Additional Details : Weight : 0.25000kg
Specifications
Tubing | |
2.54 mm | |
1000 cm | |
-50°C | |
Acids, Alkalies | |
Good |
Autoclavable | |
Tygon ST | |
For General Laboratory Applications | |
1 Each | |
Transparent | |
74°C |