Learn More
Saint Gobain Life Sciences Tygon™ E-3603 Tubing
Product Code. 10130121
Change view
Click to view available options
Diameter (English) Inner:
0.5 in.
1/4 in.
Diameter (English) Outer:
0.625 in.
1.125 in.
Maximum Pressure:
36 psi (at 73°F)
53 psi (at 73°F)
Thickness (English):
3/16 in.
5/16 in.
Unit Size:
15m
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Thickness (English) Unit Size
10130121 1/4 in. 0.625 in. 53 psi (at 73°F) 3/16 in. 15m
12355909 0.5 in. 1.125 in. 36 psi (at 73°F) 5/16 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10130121 Supplier Saint Gobain Life Sciences Supplier No. ACF00020
  • ONLINE EXCLUSIVE PRICE
    689.76€
    338.82€ / 15m
    Save 350.94€
    50% Off
View all promotions
Estimated Shipment: 14-12-2026
Tygon™ E-3603 Tubing for laboratory and vacuum applications.
Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Compliance
FDA, NSF, 3-A, meets USP Class VI, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 1/4 in.
Diameter (Metric) Inner 6.4 mm
Material Tygon S3 E-3603
Diameter (English) Outer 0.625 in.
Diameter (Metric) Outer 15.9 mm
Overall Length (English) 50 ft.
Overall Length (Metric) 15 m
Thickness (English) 3/16 in.
Thickness (Metric) 4.8 mm
Color Clear
Flexibility 13 mm bend radius
Maximum Pressure 53 psi (at 73°F)
Product Type Tubing