Learn More
Ismatec™ Tygon™ MHSL 2001 3-stop Tubing
Product Code. p-7115976
Click to view available options
Tubing Size:
0.64mm I.D., 2.44mm O.D.
1.02mm I.D., 2.72mm O.D.
1.52mm I.D., 3.22mm O.D.
2.06mm I.D., 3.76mm O.D.
2.79., I.D., 4.49mm O.D.
Unit Size:
Pack of 6
Product Code. 10387992
Brand: Ismatec™ SC0812A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Transparent, plasticizer-free 3-stop Tygon™ tubing with superior pump-life. Ismatec™ Tygon™ MHSL 2001 2-stop Tubing is especially designed for MEK and other aggressive solvents.
  • Plasticizer and oil-free.
  • Smooth inner-bore.
  • Low sorption maintains fluid and tube integrity.
  • Does not impart anything into the pumping medium.
  • No release of hazardous materials when properly incinerated.
  • Applications
    • Acids: Excellent
    • Alkaline solutions: Excellent
    • Solvents: Good/Excellent
  • Complies with: FDA.
  • Sterilization
    • Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
    • Gas sterilization with ethylene oxide.
    • Sterilization with radiation up to 2.5 mrad.
  • Note: Although designed for MS/CA cassettes, this tubing has only 2 stoppers!
Specifications
Certifications/Compliance FDA
Description Color-code: Purple-White
Autoclavable Autoclavable
Diameter (Metric) Inner 2.79 mm
Material Tygon MHSL 2001
Diameter (Metric) Outer 4.49 mm
Overall Length (Metric) 30 cm
Thickness (Metric) 0.85 mm
For Use With peristaltic pump
For Use With (Application) For MEK and other aggressive solvents. MS/CA cassettes
Min. Operating Temperature (Metric) -73°C
Packaging Quantity 6 pcs./Pk.
Resistance Properties Acids, Alkalies
Tubing Size 2.79., I.D., 4.49mm O.D.
Gas Permeability CO2 1140, O2 76, N2 190
Color Transparent
Max. Operating Temperature (Metric) 57°C
Product Type 2-stop Tubing