Learn More
Ismatec™ Tygon™ E-LFL Microbore 2-Stop Tube Sets
Click to view available options
Tubing Size:
10
12
26
34
48
Unit Size:
Pack of 12
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
- For use with Masterflex™ L/S™ multichannel pump heads
Specifications
Specifications
| Certifications/Compliance | USP; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Non-Phthalate; RoHS; Prop 65 NA (no Prop 65 chemicals) |
| Diameter (Metric) Inner | 0.89 mm |
| Material | Tygon E-LFL |
| Overall Length (English) | 1.25 ft. |
| Overall Length (Metric) | 0.38 m |
| Min. Operating Temperature (Metric) | 0°C |
| Packaging Quantity | Pk. of 12 |
| Tubing Size | 26 |
| Flow Rate | 30 mL/min |
| Maximum Pressure | 25.3 psi, 1.75 bar |
| Show More |