Learn More
Ismatec™ Tygon™ E-LFL C/L™ Microbore Pump Tubing
Product Code. p-7187330
Click to view available options
Tubing Size:
18
26
30
48
Unit Size:
Each
Product Code. 15570652
Brand: Ismatec™ 0644918
Only null left
Add to basket
Add to basket
Longest life of all Tygon™ formulations
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • For general laboratory use
  • Long life - won’t age or oxidize
  • Fits tightly to glass and metal & bends to small radius curves
  • Nontoxic
  • Transparent
Specifications
Certifications/Compliance FDA approved; Prop 65 NA (no Prop 65 chemicals)
Autoclavable Autoclavable
Diameter (Metric) Inner 0.51 mm
Material Tygon E-LFL
Overall Length (English) 100 ft.
Overall Length (Metric) 30.4 m
Min. Operating Temperature (Metric) 0°C
Tubing Size 18
Flow Rate 3.6 mL/min
Sterilization Method Autoclave, Ethylene Oxide
Maximum Pressure 25.3 psi, 1.75 bar
Max. Operating Temperature (Metric) 40°C
Product Type Microbore Tubing