Learn More
Saint Gobain Life Sciences Tygon™ 3350 Sanitary Silicone Tubing
Product Code. 12336909
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
10 mm
15 mm
18 mm
20 mm
Diameter (Metric) Outer:
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
14 mm
21 mm
24 mm
27 mm
Unit Size:
15m
Each
Product Code. 12336909
Brand: Saint Gobain Life Sciences T330L153
Only null left
Add to basket
Add to basket
Designed to reduce the risk of particle entrapment and microscopic build-up during fluid transfer. Tygon™ 3350 Sanitary Silicone Tubing features an ultra-smooth inner surface and reduced surface area for improved flow characteristics.
  • Ultra-smooth inner bore reduces potential for particle entrapment
  • Minimal extractables help maintain fluid integrity
  • Documented biocompatibility for sensitive applications
  • Excellent fluid flow characteristics
  • Fully tested to ISO 10993 standards to facilitate validation process
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001, ISO 10993
Autoclavable Autoclavable
Diameter (English) Inner 0.28 in.
Diameter (Metric) Inner 7 mm
Material Tygon 3350
Diameter (English) Outer 0.39 in.
Diameter (Metric) Outer 10 mm
Overall Length (English) 49.2 ft.
Overall Length (Metric) 15 m
Thickness (English) 0.06 in.
Thickness (Metric) 1.5 mm
Min. Operating Temperature (English) -112°F
Min. Operating Temperature (Metric) -80°C
Color Translucent
Max. Operating Temperature (English) 399.2°F
Max. Operating Temperature (Metric) 204°C
Product Type Tubing