Learn More
Saint Gobain Life Sciences Tygon™ 3350 Sanitary Silicone Tubing
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
10 mm
15 mm
18 mm
20 mm
Diameter (Metric) Outer:
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
14 mm
21 mm
24 mm
27 mm
Unit Size:
15m
Each
Description
- Ultra-smooth inner bore reduces potential for particle entrapment
- Minimal extractables help maintain fluid integrity
- Documented biocompatibility for sensitive applications
- Excellent fluid flow characteristics
- Fully tested to ISO 10993 standards to facilitate validation process
Specifications
Specifications
| Certifications/Compliance | ISO 9001, ISO 10993 |
| Autoclavable | Autoclavable |
| Diameter (English) Inner | 0.79 in. |
| Diameter (Metric) Inner | 20 mm |
| Material | Silicone |
| Diameter (English) Outer | 1.06 in. |
| Diameter (Metric) Outer | 27 mm |
| Overall Length (English) | 49.2 ft. |
| Overall Length (Metric) | 15 m |
| Thickness (English) | 0.14 in. |
| Show More |