missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 AL TF640 00
Description
- Ventilated coverall available in sizes small to 3X.
- Cuff elastication and elastic thumb loop on inner sleeve opening.
- Over boot assembly consisting of integrated dissipative inner boots attached to dissipative outerboots.
- Gray.
The protective system made of Tychem™ 6000 AL TF640 00 and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates. - The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
- The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip. In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
- Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate.
- They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Specifications
Specifications
| Color | Gray |
| For Use With (Application) | Protecting from Contaminents |
| Hood Type | Fully Encapsulated |
| Material | Tychem |
| Size | Medium |
| Closure Type | Horizontal Zipper with Double Flap Hook and Loop |
| Disposable | Single-use |
| Gender | Unisex |
| Latex Free | Yes |
| No. of Pockets | None |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?