missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 AL TF640 00
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 2
Description
- Ventilated coverall available in sizes small to 3X.
- Cuff elastication and elastic thumb loop on inner sleeve opening.
- Over boot assembly consisting of integrated dissipative inner boots attached to dissipative outerboots.
- Gray.
The protective system made of Tychem™ 6000 AL TF640 00 and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates. - The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
- The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip. In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
- Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate.
- They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Specifications
Specifications
| Color | Gray |
| For Use With (Application) | Protecting from Contaminents |
| Hood Type | Fully Encapsulated |
| Material | Tychem |
| Size | X-Large |
| Closure Type | Horizontal Zipper with Double Flap Hook and Loop |
| Disposable | Single-use |
| Gender | Unisex |
| Latex Free | Yes |
| No. of Pockets | None |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction