Learn More
DuPont™ Tychem™ 2000C Gown
Product Code. p-7228553 Shop All DuPont Products
Click to view available options
Size:
Medium
2X-Large
Unit Size:
Pack of 25
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17475512
Brand: DuPont™ D15546833
Only null left
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Shin-length gown with rear closure, knitted cuff with double cuff system, mandarin collar.
  • Double cuffs for increased protection and tighter connection with gloves
  • Elasticated wrists
  • Knitted cuff (inside) for better comfort
  • Offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
  • Used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Color Yellow
Disposable Single-use
Certifications/Compliance (EU) 2016/425, EN 14126, EN 1149-1