missing translation for 'onlineSavingsMsg'
Learn More

Benchmark™ BMC02 Limited Life Chemical Coverall - Type 5/6

Product Code. 30242990
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30242990

Brand: Benchmark™ BMC00002AP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Benchmark BMC-02 is a limited life coverall made from lightweight spun-bonded polypropylene with an outer breathable film. It is designed to protect workers from hazardous substances or sensitive products and processes from contamination.

  • SMS breathable material
  • Elasticated waist
  • Overlocked stitched seams
  • Inset sleeve for ease of movement

Specifications

Product Type Hooded Coveralls
Size 4X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Limited-use
Waist Style Loose
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Dust, Particle, Chemicals
Certifications/Compliance CE , EN 1073-2:2002 CLASS 2 , EN 13034:2005 + A1:2010 Type 6 , EN 13982-1:2004 + A1:2010 Type 5 , EN 14126:2003 + AC:2004 Type 5B,Type 6B
No. of Pockets None
For Use With (Application) Painting and Finishing, Electrical Engineering and Maintenance
Quantity 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.