Learn More
Skytec TX324L Disposable Latex-Free Nitrile Gloves
GREENER_CHOICE
Product Code. 17861084
Change view
Click to view available options
Size:
8, Medium
9, Large
10, X-Large
Unit Size:
Pack of 100
Product Code. Size unitSize
17861084 9, Large Pack of 100
17891074 8, Medium Pack of 100
17801094 10, X-Large Pack of 100
3 options
Product Code. 17861084
Brand: Skytec SKG00324IH
Only null left
Add to basket
Add to basket
TX324 is a high performance biodegradable nitrile single use examination glove. Sustainable, dual certified (Industrial and Medical) and food safe.
  • High quality nitrile, soft and durable
  • Palm thickness 0.07 mm (3 mil), weight: 3.5 g
  • Tested to ASTM D5526 (determine the degree and rate of anaerobic biodegradation of materials in accelerated landfill conditions)
    • Results: 30% biodegradation after 202 days
  • Tested to ASTM D5511 (determine the degree and rate of anaerobic biodegradation of materials in high-solids anaerobic-digestion conditions)
    • Results: 90% biodegradation after 490 days
  • Textured finish on fingertips for improved grip and touch
  • Powder-free, latex free, non-sterile, AQL 1.5
  • Chlorinated inside to offer more comfort and reduce friction when donning
  • Antistatic properties due to inherent antistatic properties of nitrile
  • Approved for food handling ((EU) No 10/2011, (EC) 1935/2004 and FDA 21 CFR 177.2600)
  • Dual labelling: PPE and medical
  • Medical device compliant (MDD 93/42/EEC), conforms to EN455 (all parts) Compliant with ASTM D6321
Specifications
Certifications/Compliance (Europe) EN374
Chlorinated Yes
Accelerator Free No
Applications Healthcare, Industrial
AQL 1.5
Color Violet
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 3 mil
Finger Thickness (Metric) 0.07 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3 mil
Palm Thickness (Metric) 0.07 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 9, Large
Sterility Non-sterile
Thickness (English) 3 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Series TX 324L
Length (English) 9 in.
Length (Metric) 24 cm