Learn More
Thermo Scientific™ TX-750 Swinging Bucket/BIOShield 1000A Adapters
Product Code. p-4527797 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
1.5/2.0 mL Conical Microtube
10 mL Blood Collection/15mL DIN Tube
100 mL Round-bottom Open-top Tube
14 mL Flanged Round-bottom Tube
15 mL Conical Urine Tube
150 mL Round-bottom Open-top Tube
25 mL DIN Round-bottom Tube
25 mL Universal Tube
250 mL Flat-bottom Bottles
45 mL Flat/Round Tube
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube
50 mL Round-bottom Tube
Unit Size:
Pack of 4
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11874471 250 mL Flat-bottom Bottles Pack of 4
11884471 150 mL Round-bottom Open-top Tube Pack of 4
11894471 100 mL Round-bottom Open-top Tube Pack of 4
11814481 50 mL Round-bottom Tube Pack of 4
11824481 45 mL Flat/Round Tube Pack of 4
11834481 25 mL Universal Tube Pack of 4
11844481 25 mL DIN Round-bottom Tube Pack of 4
11884501 14 mL Flanged Round-bottom Tube Pack of 4
11864481 15 mL Conical Urine Tube Pack of 4
11894501 10 mL Blood Collection/15mL DIN Tube Pack of 4
11874481 5/7 mL Blood Collection Tube Pack of 4
11884481 5/7 mL Round-bottom Tube Pack of 4
11894481 1.5/2.0 mL Conical Microtube Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11874471 Supplier Thermo Scientific™ Supplier No. 75003737
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
  • Maximum tube size: 63 × 130mm
TRUSTED_SUSTAINABILITY
Specifications
Type Swinging Bucket Adapter
Capacity 250 mL Flat-bottom Bottles