Learn More
Masterflex™ Two-way Normally Closed Solenoid Pinch Valve
Description
- Ideal where contamination is an issue
- Control flow in your tubing without direct contact between the fluid and the valve
- Fast response time
- Low-power consumption and high-cycle life
Specifications
Specifications
| Description | 12 VDC |
| Material | Silicone |
| For Use With | 1/32 in. ID x 3/32 in. OD tubing |
| For Use With (Application) | Excellent for sanitary and pharmaceutical applications |
| Packaging Quantity | 1 Pack |
| Maximum Pressure | 15 psi |
| Product Type | Solenoid Electric Valves (actuated) |