Learn More
Köttermann Two Door Tall Storage Cabinet
Product Code. 17392089
Change view
Click to view available options
Door Type:
Hinged
Sliding
No. of Shelves:
4
None
Dimensions (D x W x H):
350 x 900 x 1920 mm
366 x 900 x 1920 mm
500 x 900 x 1920 mm
516 x 900 x 1920 mm
350 x 1200 x 1920 mm
500 x 1200 x 1920 mm
770 x 1100 x 2250 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Type No. of Shelves Dimensions (D x W x H) Unit Size
17392089 Sliding 4 500 x 900 x 1920 mm Each
17362089 Sliding 4 350 x 900 x 1920 mm Each
17342079 Sliding 4 350 x 1200 x 1920 mm Each
17382089 Hinged 4 366 x 900 x 1920 mm Each
17362079 Sliding 4 500 x 1200 x 1920 mm Each
17312099 Hinged 4 516 x 900 x 1920 mm Each
17322099 Hinged None 770 x 1100 x 2250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17392089 Supplier Köttermann Supplier No. 30100077
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
This Two Door Tall Storage Cabinet features a two hinged or sliding doors to protect stored equipment and utensils.
  • Tall, slender design expands storage with a small footprint
  • Two doors protect stored items
  • Sliding and hinged door options available
Specifica
Color White
Adjustable No
Cabinet Type Tall Storage
Dimensions (D x W x H) 500 x 900 x 1920 mm
Door Type Sliding
Finish Epoxy Coated
For Use With (Application) Storage
No. of Doors 2
No. of Drawers None
No. of Shelves 4
Warranty None
Weight (Metric) 99.4 kg
With Casters No
Material Steel
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.