Learn More
Brannan™ Twaddell Hydrometers
Product Code. 12869207
Click to view available options
Specific Gravity Range:
102 to 138 g/mL
24 to 48 g/mL
48 to 74 g/mL
74 to 102 g/mL
Unit Size:
Each
Product Code. 12869207
Brand: Brannan™ 63/100/1
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Shop loaded Twaddell hydrometers for specific gravity range 24 to 48g/mL
For 0 to 24 Twaddell range subdivided 1.0TW. 158mm Shot loaded, scale subdivision 1.0TW, accuracy ±1.0TW, 0.5TW ±0.5TW at 60/60°F. Calibrated at 15.6°C.
Specifications
Quantity 1Pack
Type Twaddle Hydrometer
Accuracy ±1Tw
Increments 1.0TW
Length (Metric) 250 mm
Specific Gravity Range 74 to 102 g/mL