Learn More
IKA™ TW.CABLE Connection Cable
Product Code. 17775868 Shop All IKA Products
Click to view available options
Length (Metric):
0.5 m
Unit Size:
Each
Product Code. 17775868
Brand: IKA™ 0020025856
Only null left
Add to basket
Add to basket
The TW.CABLE connection cable can be used to electrically connect separate clusters of MIXSTER devices.
  • Clusters require only one power supply in total
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) IKA™ MIXSTER Universal and Compact Mixing Devices
Length (Metric) 0.5 m