missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas TV400K Cooled Twin-Plate Maxi-Gel Vertical Electrophoresis Unit
Description
- Twin-plate 20.5 x 20cm maxi-gel unit
- Large format with 20.5 x 20cm glass plate dimensions are ideal for running higher percentage polyacrylamide gels to resolve smaller proteins of similar MW
- A cooling coil within the gel tank allows faster separations to be performed at high voltage without compromising resolution
- Snap-lock connectors allow the cooling coil to be connected to an external chiller unit for faster, higher voltage separations
- Float glass plates of 4mm thick provides uniform gel thickness and sample migration, and with ground edges inhibits sample leakage during casting
- Triple screws present on each gel-clamp acts in conjunction with a counterbalanced, dovetailed silicone gasket to disperse pressure evenly along the height and breadth of the gel as the screws are tightened
Specifications
Specifications
| Cooling Type | Cooling coil connected to an external chiller unit |
| Running Time | 2 to 2.5 hr. |
| Type | TV400K Cooled Twin-Plate Maxi-Gel Vertical Electrophoresis Unit |
| Gel Dimensions | W x H: 16.5 x 17.5 cm |
| Includes | Twin-plate wide format mini-gel unit with GRM and cooled gel tank, with built-in cooling coil and quick-fit tubing, plus lid, 2 x (20.5W x 20cmH) plain glass plates, 4 x 1mm spacers, 2 x spacer aligners, 2 x (20.5 x 20cm) notched glass plates, 1 x dummy plate and 2 x 1mm thick 24-sample combs |
| Thickness (Metric) | Plate: 0.4 cm |
| Thickness (Metric) Spacer | 0.1 cm |
| Volume (Metric) | Gel Tank: 4200 mL |
| Volume (Metric) Buffer | Inner Buffer Chamber: 700 mL |
| Dimensions (L x W x H) | W x H: 38 x 27.5 cm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?