missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas TV200 Standard Twin-Plate Wide Format Mini-Gel Electrophoresis Unit
Description
- Twin-plate 20.5W x 10cmH wide format mini-gel unit
- Designed specifically for PAGE techniques requiring self-cast gels
- Wide format of 20.5cm gel width effectively allows two mini-gels to be run within the same gel
- Glass plate stops within the newly modified GRM locks the glass plates firmly into position onto soft silicone gaskets in the casting base to ensure leak-free gel-casting
- Float glass plates of 4mm thick provides uniform gel thickness, sample migration and ground edges inhibits sample leakage during casting
- Twin screws on each gel-clamp acts in conjunction with a counterbalanced, dovetailed silicone gasket to disperse pressure evenly along the height and breadth of the gel as the screws are tightened
Specifications
Specifications
| Running Time | 1.5 to 2 hr. |
| Type | TV200 Standard Twin-Plate Wide Format Mini-Gel Electrophoresis Unit |
| Gel Dimensions | W x H: 16.5 x 8.5 cm |
| Includes | Twin-plate wide format mini-gel unit with GRM and gel tank, lid, 2 x (20.5W x 10cmH) plain glass plates, 4 x 1mm spacers, 2 x spacer aligners, 2 x (20.5 x 10cm) notched glass plates, 1 x dummy plate and 2 x 1mm thick 24-sample combs |
| Thickness (Metric) | Plate: 0.4 cm |
| Thickness (Metric) Spacer | 0.1 cm |
| Volume (Metric) | Gel Tank: 2800 mL |
| Volume (Metric) Buffer | Inner Buffer Chamber: 300 mL |
| Dimensions (L x W x H) | W x H: 30 x 14 cm |
| Depth (Metric) | 15 cm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?