Learn More
Honeywell™ Turboshield Visor
Description
- Feel safer with 50% more chin coverage compared to traditional visors, without the need for bulky accessories
- Visors can be easily removed with the new Turboshield Faceshield system using a simple push-button release.
Spécification
Spécification
| Certifications/Compliance | EC Category PPE: 2, EC Certificate: C1519.2HON, EC Attestation: C1519.2HON, EN 166, EN 169, EN 170, EN 172 |
| Lens Coating | Anti-fog |
| Description | Dielectric |
| Lens Tint | Clear |
| For Use With (Equipment) | Turboshield headgear |
| For Use With (Application) | Suitable face protection for most indoor work applications |
| Lens Material | Polycarbonate |