Learn More
Honeywell™ Turboshield Headgear
Click to view available options
For use with Turboshield face shields
Description
- Flexible head-cushioning suspension cradle provides 300% greater contact area than traditional suspensions to eliminate pressure points for all-day wear
- Simple push-button release system allows for easy visor exchange in seconds
- Control knobs are large and ergonomically designed for easy adjustment - even while wearing gloves
- Breathable, removable, washable headband
- Smooth-pivoting action with positive stops in raised and lowered position
Specifications
Certifications/Compliance | EC Category PPE: 2, EC Certificate: C1519.2HON, EC Attestation: C1519.2HON, EN 166 |
Description | Dielectric |
For Use With (Equipment) | Turboshield visor face shield |
For Use With (Application) | Impact protection, splash protection, UV protection |
Frame Color | Gray |
No. per Pack | 1 Ea. |
Frame Material | Browguard is ABS, Suspension is Nylon 66 |