Learn More
WTW™ Turb™ Calibration Kits
Product Code. p-8002866
Click to view available options
For Use With (Equipment):
Turb 350 IR Portable Turbidimeter
Turb 355 T, 335 IR Portable Turbidimeter
Unit Size:
Each
Product Code. 10033022
Brand: WTW™ 600541
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Get optimum calibration results for your turbidimeters with the Turb™ calibration kits.
  • Includes cleaning tissue and 4 indexing rings
  • For use with Turb&reg 350 IR Portable turbidimeter
  • Turbidity of 0.02 - 10 - 100 - 1000NTU
  • Calibration set
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Turb 350 IR Portable Turbidimeter
Description Calibration kit
Includes Cleaning tissue and 4 indexing rings
Turbidity 0.02, 10, 100, 1000 NTU