Learn More
RETSCH Tungsten Carbide Grinding Balls
Shop All Retsch Products
Click to view available options
For Use With (Equipment):
MM 200 and MM 301
MM 200 and MM 400
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
3 mm
4 mm
5 mm
7 mm
10 mm
12 mm
15 mm
20 mm
30 mm
40 mm
Unit Size:
Each
Pack of 200
Specifications
Specifications
| Product Type | Grinding Balls |
| Diameter (Metric) | 5 mm |
| For Use With (Equipment) | Reaction Vials |
| Material | Tungsten Carbide |