Learn More
Masterflex™ Tubing adapter NPT (M) PVDF
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
177.00€
Specifications
Product Type | Vacuum Tubing |
---|---|
For Use With | Masterflex L/S and I/P sanitary tubing connection |
Outlet Port | Sanitary x NPT(M) |
Material (Seal) | PVDF |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
12498670
|
Masterflex™
31201-01 |
1/4 in. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
12488680
|
Masterflex™
31201-03 |
1/2 in. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Determine which connector type is needed for your application by contacting free technical application assistance
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most materials and configurations available to ship same day
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
- All fittings are compatible with Tri-Clamp™ connections
- Excellent chemical resistance; low leachable characteristics
Specifications
Vacuum Tubing | |
Masterflex L/S and I/P sanitary tubing connection | |
Sanitary x NPT(M) | |
PVDF |