Learn More
Thermo Scientific™ Tubes, strips of 8, red
Click to view available options
Quantity:
250 strips
Unit Size:
Pack of 250
Description
- Compatible with 0.2 mL thermal cycler blocks
- Sold without caps
- Compatible with 0.2 mL thermal cycler blocks
- 8 tube strips
- Colored tubes are ideal for keeping track of samples
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
- Applications PCR, qPCR/Real-Time PCR
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Color | Red |
| Volume (Metric) Working | 0.2 mL |
| Volume (Metric) | 0.25 mL |
| Quantity | 250 strips |
| For Use With (Equipment) | Thermal Cyclers |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Reaction Speed | Standard |
| Closure Type | Not Included |
| Show More |