Learn More
Greiner Bio-One Tubes
Product Code. 10547091
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
28 mL
missing translation for 'unitSize'
Pack of 1200
Product Code. 10547091
missing translation for 'mfr': Greiner Bio-One 205111
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Diameter (Metric) 26 mm
Product Type Tube
Capacity (Metric) 28 mL
No. per Case 1200
Height (Metric) 64 mm