Learn More
Carbolite™ Tube Furnace
Product Code. 12029159
Change view
Click to view available options
Depth (Metric):
175 mm
332 mm
375 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) unitSize
12029159 375 mm Each
12099149 175 mm Each
12029089 332 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12029159 Supplier Carbolite™ Supplier No. M1225250230SN+&023508P1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Analysis Time 15 min.
Type Tube Furnace
Includes Standard Controller
Temperature Stability 1200°C
Dimensions (L x W x H) Exterior 370 W x 375 H mm
Length (Metric) 300 mm
Width (Metric) 370 mm
Height (Metric) 375 mm
Depth (Metric) 375 mm