Learn More
behr Labor-Technik™ Tube Cyanide Trapping Valve
Product Code. 10074741
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10074741 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10074741 Supplier behr Labor-Technik™ Supplier No. B00217963
Absorption vessel ASH/CY complete, with center connection MV 2914 and ground joint clamp SKL 14 and glass pearl TL 1
Specifications
For Use With (Application) For Cyanide
Type Tube Cyanide Trapping Valve
Quantity 1