Learn More
SCAT™ Tube Connector, Straight
Product Code. 10285063 Shop All SCAT Europe Products
Change view
Click to view available options
Diameter (Metric):
5 to 7 mm
6 to 8 mm
6.2 to 7.5 mm
9.5 to 10 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10285063 9.5 to 10 mm Each
10024257 6.2 to 7.5 mm Each
10349637 5 to 7 mm Each
10631021 6 to 8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10285063 Supplier SCAT™ Supplier No. 107817
  • ONLINE EXCLUSIVE PRICE
    10.95€
    10.05€ / Pack of 1
    Save 0.90€
    8% Off
View all promotions
Estimated Shipment: 16-10-2026
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Diameter (Metric) 9.5 to 10 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing
For Use With (Application) HPLC/UHPLC Systems
Color Transparent
Product Type Tubing Connector