Learn More
Julabo™ Tube Clamps for Baths
Click to view available options
Holds:
0.75 in. I.D. Reinforced tubing
1 in. I.D. Reinforced tubing
12mm /0.5 in. ID. Reinf. tubing
8mm I.D.CR tubing
CR 10 mm ID, Reinf. 8 mm ID tubing
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | CF30, CF31, CF40, CF41 Cryo-compact circulator, ED-13 open heating bath circulator |
| Type | 2 Tube Clamps |
| Holds | 8mm I.D.CR tubing |
| Size | 1 |