Learn More
Socorex PTFE delivery Jet Pen tubing, 60 cm for bottle top
Brand:  Socorex 1.524
PTFE delivery Jet Pen tubing, 60 cm for bottle top
Product Code. 10492925
  • ONLINE EXCLUSIVE PRICE
    112.00€
    86.40€ / Each
    Save 25.60€
    22% Off