Learn More
TTC33 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558314
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EQQKVAQRIK KSEAPAEVTH FSPKSIPDYD FESDEIVAVC AAIAEKEKTV SANKTMVIVS ASGAIETVTE KEDGATPPDG SV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.
TTC33 is a protein coding gene.Specifications
TTC33 | |
Polyclonal | |
Unconjugated | |
Ttc33 | |
2410099M07Rik; 2900001O04Rik; AI507072; AW538342; osmosis responsive factor; osmosis-responsive factor; OSRF; RGD1564042; tetratricopeptide repeat domain 33; tetratricopeptide repeat protein 33; TPR repeat protein 33; TTC33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23548 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PID6 | |
Ttc33 | |
Recombinant protein corresponding to Human TTC33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only