missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSC Selective Agar

Product Code. 11993762
missing translation for 'orderingAttributeHoverText'
Quantity:
10 Plates
missing translation for 'unitSize'
Pack of 10
This item is not returnable. View return policy

Product Code. 11993762

missing translation for 'mfr': Thermo Scientific™ PO0520A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enhance the detection of Clostridium species with this agar, offering superior selectivity and differentiation for clinical and environmental samples.

Enhance the detection of Clostridium species with TSC Selective Agar, offering superior selectivity and differentiation for clinical and environmental samples.

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.