Learn More
Tryptanthrin, TRC
Specifications
Specifications
| Chemical Name or Material | Tryptanthrin |
| CAS | 13220-57-0 |
| Molecular Formula | C15H8N2O2 |
| Quantity | 25 mg |
| Synonym | Indolo[2,1-b]quinazoline-6,12-dione, Couroupitine A, NSC 349447, TryptanthrinTryptanthrine |
| InChI Formula | InChI=1S/C15H8N2O2/c18-13-10-6-2-4-8-12(10)17-14(13)16-11-7-3-1-5-9(11)15(17)19/h1-8H |
| SMILES | O=C1N2C(=Nc3ccccc13)C(=O)c4ccccc24 |
| IUPAC Name | indolo[2,1-b]quinazoline-6,12-dione |
| Molecular Weight (g/mol) | 248.24 |
| Formula Weight | 248.0586 |