missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Trypsin Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Concentration | 100 mg/mL |
| For Use With (Application) | ELISA |
| Quantity | 500 mg |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Reconstitution | Reconstitute in sterile distilled water. |
| Protein | Trypsin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction