Learn More
Trypan Blue Monoazo Impurity Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Trypan Blue Monoazo Impurity Sodium Salt |
| CAS | 29642-16-8 |
| Molecular Formula | C24H21N3O7S2.2Na |
| Quantity | 25 mg |
| Synonym | Trypan Blue Impurity 1 |
| InChI Formula | InChI=1S/C24H21N3O7S2.2Na/c1-13-4-3-5-15(8-13)16-6-7-20(14(2)9-16)26-27-23-21(36(32,33)34)11-17-10-18(35(29,30)31)12-19(25)22(17)24(23)28;;/h3-12,28H,25H2,1-2H3,(H,29,30,31)(H,32,33,34);; |
| SMILES | CC1=CC=CC(C2=CC=C(C(C)=C2)/N=N/C3=C(C4=C(C=C(S(=O)(O)=O)C=C4C=C3S(=O)(O)=O)N)O)=C1 |
| IUPAC Name | 5-amino-3-((3,3'-dimethyl-[1,1'-biphenyl]-4-yl)diazenyl)-4-hydroxynaphthalene-2,7-disulfonic acid','disodium salt |
| Molecular Weight (g/mol) | 573.55 |
| Formula Weight | 573.06 |