Learn More
Trypan Blue Monoazo Impurity Sodium Salt, TRC
Product Code. 30411500
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411500 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30411500 Supplier Toronto Research Chemicals Supplier No. TRCT22389050MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trypan Blue Monoazo Impurity Sodium Salt
CAS 29642-16-8
Molecular Formula C24H21N3O7S2.2Na
Quantity 50 mg
Synonym Trypan Blue Impurity 1
InChI Formula InChI=1S/C24H21N3O7S2.2Na/c1-13-4-3-5-15(8-13)16-6-7-20(14(2)9-16)26-27-23-21(36(32,33)34)11-17-10-18(35(29,30)31)12-19(25)22(17)24(23)28;;/h3-12,28H,25H2,1-2H3,(H,29,30,31)(H,32,33,34);;
SMILES CC1=CC=CC(C2=CC=C(C(C)=C2)/N=N/C3=C(C4=C(C=C(S(=O)(O)=O)C=C4C=C3S(=O)(O)=O)N)O)=C1
IUPAC Name 5-amino-3-((3,3'-dimethyl-[1,1'-biphenyl]-4-yl)diazenyl)-4-hydroxynaphthalene-2,7-disulfonic acid','disodium salt
Molecular Weight (g/mol) 573.55
Formula Weight 573.06