missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trovafloxacin Mesylate, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Trovafloxacin Mesylate |
| CAS | 147059-75-4 |
| Molecular Formula | C20 H15 F3 N4 O3 . C H4 O3 S |
| Quantity | 1 mg |
| Synonym | 1,8-Naphthyridine-3-carboxylic acid','7-[(1alpha,5alpha,6alpha)-6-amino-3-azabicyclo[3.1.0]hex-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-','methanesulfonate (1:1), 1,8-Naphthyridine-3-carboxylic acid','7-(6-amino-3-azabicyclo[3.1.0]hex-3-yl)-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-','(1alpha,5alpha,6alpha)-','monomethanesulfonate (9CI), 3-Azabicyclo[3.1.0]hexane','1,8-naphthyridine-3-carboxylic acid deriv., CP 99219, CP 99219-27, Trovafloxacin mesylate, Trovafloxacin methanesulfonate, Trovafloxacin monomethanesulfonate, Trovan, 1,8-Naphthyridine-3-carboxylic acid','7-[(1α,5α,6α)-6-amino-3-azabicyclo[3.1.0]hex-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-','methanesulfonate (1:1) |
| InChI Formula | InChI=1S/C20H15F3N4O3.CH4O3S/c21-8-1-2-15(13(22)3-8)27-7-12(20(29)30)17(28)9-4-14(23)19(25-18(9)27)26-5-10-11(6-26)16(10)24;1-5(2,3)4/h1-4,7,10-11,16H,5-6,24H2,(H,29,30);1H3,(H,2,3,4)/t10-,11+,16+; |
| SMILES | CS(=O)(=O)O.N[C@@H]1[C@H]2CN(C[C@@H]12)c3nc4N(C=C(C(=O)O)C(=O)c4cc3F)c5ccc(F)cc5F |
| IUPAC Name | 7-[(1S,5R)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid |
| Molecular Weight (g/mol) | 512.46 |
| Formula Weight | 512.0977 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction