missing translation for 'onlineSavingsMsg'
Learn More

Trovafloxacin Mesylate, TRC

Product Code. 30453301
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30453301

Brand: Toronto Research Chemicals TRCT8930005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trovafloxacin Mesylate
CAS 147059-75-4
Molecular Formula C20 H15 F3 N4 O3 . C H4 O3 S
Quantity 5 mg
Synonym 1,8-Naphthyridine-3-carboxylic acid','7-[(1alpha,5alpha,6alpha)-6-amino-3-azabicyclo[3.1.0]hex-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-','methanesulfonate (1:1), 1,8-Naphthyridine-3-carboxylic acid','7-(6-amino-3-azabicyclo[3.1.0]hex-3-yl)-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-','(1alpha,5alpha,6alpha)-','monomethanesulfonate (9CI), 3-Azabicyclo[3.1.0]hexane','1,8-naphthyridine-3-carboxylic acid deriv., CP 99219, CP 99219-27, Trovafloxacin mesylate, Trovafloxacin methanesulfonate, Trovafloxacin monomethanesulfonate, Trovan, 1,8-Naphthyridine-3-carboxylic acid','7-[(1α,5α,6α)-6-amino-3-azabicyclo[3.1.0]hex-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-','methanesulfonate (1:1)
InChI Formula InChI=1S/C20H15F3N4O3.CH4O3S/c21-8-1-2-15(13(22)3-8)27-7-12(20(29)30)17(28)9-4-14(23)19(25-18(9)27)26-5-10-11(6-26)16(10)24;1-5(2,3)4/h1-4,7,10-11,16H,5-6,24H2,(H,29,30);1H3,(H,2,3,4)/t10-,11+,16+;
SMILES CS(=O)(=O)O.N[C@@H]1[C@H]2CN(C[C@@H]12)c3nc4N(C=C(C(=O)O)C(=O)c4cc3F)c5ccc(F)cc5F
IUPAC Name 7-[(1S,5R)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid
Molecular Weight (g/mol) 512.46
Formula Weight 512.0977
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.