Learn More
Testo™ PTFE Thermocouple
Click to view available options
Response Time:
5 sec.
Unit Size:
Each
Specifications
Specifications
| Includes | Thermocouple, 1500mm L Cable, 1.5mm dia. Probe Shaft, PTFE, –50 to 250°C Measuring Range, 5 sec. Reaction Time, Flexible, With TC Adapter |
| Certifications/Compliance | EN 60584-2 |
| Material | PTFE |
| Response Time | 5 sec. |