Learn More
Tropine-3-thiol Hydrochloride, TRC
Product Code. 30451904
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451904 1 g -
30449189 25 g 25g
30450419 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30451904 Supplier Toronto Research Chemicals Supplier No. TRCT8926751G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 908266-48-8
Molecular Formula C8H16ClNS
Molecular Weight (g/mol) 193.74
Synonym (3-endo)-8-Methyl-8-Azabicyclo[3.2.1]octane-3-thiol Hydrochloride
IUPAC Name (5S)-8-methyl-8-azabicyclo[3.2.1]octane-3-thiol;hydrochloride
SMILES Cl.CN1[C@H]2CCC1C[C@H](S)C2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H15NS.ClH/c1-9-6-2-3-7(9)5-8(10)4-6;/h6-8,10H,2-5H2,1H3;1H/t6-,7?,8+;/m0./s1
Formula Weight 193.0692
Chemical Name or Material Tropine-3-thiol Hydrochloride