Learn More
Motic Fluorescence Filter Set for BA410E/AE31E
Product Code. 13228169
Click to view available options
Product Line:
DAPI/Hoechst/AlexaFluor 350
TRITC/Cy3/TagRFP/AlexaFluor 546
Unit Size:
Each
Product Code. 13228169
Brand: Motic 1101000203291
Only null left
Add to basket
Add to basket
This is an accessory part for the BA410E series and AE31E series. Filter sets require an EPI-Fluorescence attachment for BA or attachment for AE to function.
    li>Valid for BA410E Binocular Sextuple 50W, BA410E Binocular Sextuple 100W, BA410E Trinocular Sextuple 50W, BA410E Trinocular Sextuple 100W, AE31E Binocular, AE31E Trinocular, AE31E Binocular 100W, AE31E Trinocular 100W
Specifications
Type Filter Cube
Product Line TRITC/Cy3/TagRFP/AlexaFluor 546