missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane, Research Grade, Serva Electrophoresis™

Product Code. 11862090
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11862090 250 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11862090 Supplier Serva Electrophoresis Supplier No. 3719001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 77-86-1
Melting Point 170°C
Color White
Density 1.35g/cm3
pH 10 to 11.5
Molecular Formula C4H11NO3
Quantity 250 g
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, trizma, 2-amino-2-hydroxymethyl-1,3-propanediol, 2-amino-2-hydroxymethyl propane-1,3-diol, tris buffer
Solubility Information 800g/L
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Molecular Weight (g/mol) 121.14
PubChem CID 6503
ChEBI CHEBI:9754
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.