missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyethyl)orthoborate (>90%), TRC

Product Code. 30453462
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453462 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453462 Supplier Toronto Research Chemicals Supplier No. TRCT8879031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(2-methoxyethyl)orthoborate
CAS 14983-42-7
Molecular Formula C9H21BO6
Quantity 1 g
Synonym Tris(2-methoxyethoxy)borane, Tris(2-methoxyethyl) Borate
InChI Formula InChI=1S/C9H21BO6/c1-11-4-7-14-10(15-8-5-12-2)16-9-6-13-3/h4-9H2,1-3H3
SMILES B(OCCOC)(OCCOC)OCCOC
IUPAC Name tris(2-methoxyethyl) borate
Molecular Weight (g/mol) 236.07
Formula Weight 236.1431
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.