Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 365D0321
Additional Details : Weight : 0.20000kg
Description
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
Splitless and OC | |
Fixed Needle Syringe | |
85 mm |
26s G | |
10 μL | |
Cone |