Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
Product Code. 13406856 Shop All Thermo Scientific Products
Change view
Click to view available options
Needle Gauge:
23s G
26s G
Length (Metric) Needle:
57 mm
85 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (Metric) Needle Unit Size
13406856 23s G 57 mm Each
13436856 26s G 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13406856 Supplier Thermo Scientific™ Supplier No. 365D0271
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Edge
Add to basket
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Description Merlin Valve Injector, PTV and Split
Needle Gauge 23s G
Product Type GC Autosampler Syringe, Fixed Needle
Volume (Metric) 10 μL
Length (Metric) Needle 57 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus RSH Autosampler and TriPlus 100LS Autosampler