Learn More
BTX™ Triple Electroporation Tweezertrode
Product Code. 15741078
Change view
Click to view available options
Diameter (Metric):
3 mm
5 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15741078 3 mm Each
15625028 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15741078 Supplier BTX™ Supplier No. 450493
  • ONLINE EXCLUSIVE PRICE
    5685.00€
    4882.02€ / Pack of 1
    Save 802.98€
    14% Off
Estimated Shipment: 14-12-2026
Triple Electroporation Tweezertrode is a reusable non-invasive, tweezer-style electrode for drug or gene delivery in animal tissues.
Triple Tweezertrodes have:
  • Three electrode contacts with adjustable position and polarities
  • Improves electrical field distribution for in utero electroporation applications
Compatability:
  • ECM830
  • ECM2001+
  • Gemini X2
  • Connect to BTX generators using the Tweezertrode Connection cables (45-0204)
Applications:
  • In vivo drug or gene delivery
  • Ex vivo drug or gene delivery
  • In utero drug or gene delivery
Tweezertrodes may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
Specifications
Autoclavable Not Autoclavable
Type Tweezers
Disposable Reusable
For Use With (Equipment) ECM 830, ECM 2001, Gemini X2
Sterility Non-sterile
Temperature (Metric) Range 5°C to 40°C
Diameter (English) 0.12 in.
Diameter (Metric) 3 mm
Electrical Requirements 0 to 200 VDC (Do Not Use AC Current)